Triangular Tungsten Carbide Knife, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71019-10
103306-272EA 875.12 USD
103306-272
Triangular Tungsten Carbide Knife, Electron Microscopy Sciences
Knives
These knives will fit most any ultramicrotomes and take the place of using glass knives
  • Section thickness: 2 to 15 microns
Perfect for trimming blocks, block facing and for sectioning hard samples. The quality does compare to glass knives and the blades can be resharpened if desired.