Brain Punch Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
57400 57391 57393 57395 57397 57399 57403 57404 57405 57401 57402
103303-970EA 293.74 USD
103303-970 103305-072 103305-074 103305-076 103305-078 103305-080 103305-470 103305-472 103305-474 103305-792 103306-266
Brain Punch Kit, Electron Microscopy Sciences
Biopsy Punches
The Brain Punch Kit is a set of electropolished and sharpened punches for the Palkovit's punch technique.
Each punch has an electro polished sharp end for smooth cuts and minimal tissue adhesion. The smaller tubes are reinforced by an outer layer of concentric tubing except at the tip.