Recessed SEM Clamp, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75954-05
103305-212EA 197.24 USD
103305-212
Recessed SEM Clamp, Electron Microscopy Sciences
Clamps General Purpose Clamps
Achieve background-free imaging and no contact with sample surface in imaging area with this 5 mm recessed clip.
  • Clamping area is 25 mm (1") long x 15 mm (0.6") wide
  • Maximum thickness is 7 mm under the clamping strips
  • Overall Dimensions: 33×25×11 mm (1.3×1×0.43")
  • Made of Machined aluminum with brass screws