Measure-Matic™ Measuring Dispenser, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64299-08
103302-510EA 60.17 USD
103302-510
Measure-Matic™ Measuring Dispenser, Electron Microscopy Sciences
Bottle Top Dispensers
Makes repeated measuring and pouring easy!
  • Adjust the dispensing cup to the desired volume, then squeeze the 500 mL (16 oz) reservoir bottle
  • The desired volume of liquid will flow through the tube and into the cup
  • Excess liquid will flow back into the reservoir bottle
  • Cup and closure are made of rugged polypropylene
  • Reservoir bottle is made of squeezable-density polyethylene
Adjustable for volumes from 5 mL to 25 mL. Just squeeze and check the fluid level. Dust cover included.
Delivery information: Dust cover included.