ChillDown Silver Bioguard Anti-Microbial Floor Mats, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77519-1
103302-396EA 41.98 USD
103302-396 103302-496 103302-536
ChillDown Silver Bioguard Anti-Microbial Floor Mats, Electron Microscopy Sciences
Floor Mats Traction and Anti-Fatigue Floor Mats
These mats have nanoscale silver particles which bind with proteins to neutralize 99.9% of common microbes.
  • Minimize workspace contamination
  • Thickness less than 1 mm
  • Backed with anti-skid coating
  • Perfect for benchtop and culture hood
These mats provide better results by ensuring consistant sample reproduction. Silver is used extensively in medical applications where microbial contamination is a concern.