Small FIB Grid Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75950-02
103302-224EA 112.5 USD
103302-224
Small FIB Grid Holder, Electron Microscopy Sciences
Tools Storage
Handy grid holder which holds up to two FIB grids of the same thickness for focused ion beam.
  • Vacuum grade aluminum with brass screws
  • Also be used to safely store FIB grids with thin sections attached
Overall dimensions: 12.7 mm (1/2") x 7.8 mm (0.3 mm) H without pin
Pin: Standard 3.2 mm (1/8")
Delivery information: Includes Philips screwdriver.