Anti-DNA Ligase I Mouse Monoclonal Antibody [clone: 10H5]

Supplier: Novus Biologicals
NB100-119
103294-312EA 635.51 USD
103294-312
Anti-DNA Ligase I Mouse Monoclonal Antibody [clone: 10H5]
Antibodies
The DNA Ligase I Antibody (10H5) from Novus Biologicals is a mouse monoclonal antibody to DNA Ligase I. This antibody reacts with human, mouse. The DNA Ligase I Antibody (10H5) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Type: Primary
Antigen: DNA Ligase I
Clonality: Monoclonal
Clone: 10H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR