Micronic Push Cap Capper, CP400, Nova Biostorage Plus
Supplier: Nova Biostorage Plus
MP35050
103289-372EA 3413.08 USD
103289-372
Micronic Push Cap Capper, CP400, Nova Biostorage Plus
Closures Test/Sample Tube Closures
Micronic push cap capper CP400 is a cost effective way to seal 96 push cap tubes at once. Built to meet the varying needs of different users, the micronic push cap capper CP400 can seal the complete range of micronic TPE or EVA style capmats, capbands and capclusters.
  • Compatibility - CP400 can cap tubes (upto 1.40 ml) in ANSI/SLAS format racks up to 48 mm in height
  • User-friendly - CP400 is a compact and easy to use unit that doesn’t require installation
  • Custom Adapters - CP400 custom adapter helps to fit the height of the specific tubes being used
The capper is also compatible with other brands of storage tubes and racks.
Delivery information: The CP400 is supplied with a custom adapter.