Anti-ZNF766 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-92643
103284-680EA 706.28 USD
103284-680
Anti-ZNF766 Rabbit Polyclonal Antibody
Antibodies
The ZNF766 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ZNF766. This antibody reacts with human. The ZNF766 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: ZNF766
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR