Anti-pre-mRNA cleavage factor I (59 kDa subunit) Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-89868
103279-932EA 706.28 USD
103279-932
Anti-pre-mRNA cleavage factor I (59 kDa subunit) Rabbit Polyclonal Antibody
Antibodies
The pre-mRNA cleavage factor I (59 kDa subunit) Antibody from Novus Biologicals is a rabbit polyclonal antibody to pre-mRNA cleavage factor I (59 kDa subunit). This antibody reacts with human, mouse, rat. The pre-mRNA cleavage factor I (59 kDa subunit) Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: pre-mRNA cleavage factor I (59 kDa subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR