Anti-POLR2A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-87785
103270-804EA 748.74 USD
103270-804
Anti-POLR2A Rabbit Polyclonal Antibody
Antibodies
The RNA Polymerase II / POLR2A Antibody from Novus Biologicals is a rabbit polyclonal antibody to RNA Polymerase II / POLR2A. This antibody reacts with human. The RNA Polymerase II / POLR2A Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: POLR2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR