Anti-DNA Ligase III Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-87720
103270-768EA 767.69 USD
103270-768
Anti-DNA Ligase III Rabbit Polyclonal Antibody
Antibodies
The DNA Ligase III Antibody from Novus Biologicals is a rabbit polyclonal antibody to DNA Ligase III. This antibody reacts with human, mouse, rat. The DNA Ligase III Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: DNA Ligase III
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR