Anti-C4orf21/prematurely terminated mRNA decay factor-like Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-83750
103268-090EA 706.28 USD
103268-090
Anti-C4orf21/prematurely terminated mRNA decay factor-like Rabbit Polyclonal Antibody
Antibodies
The C4orf21 / prematurely terminated mRNA decay factor-like Antibody from Novus Biologicals is a rabbit polyclonal antibody to C4orf21 / prematurely terminated mRNA decay factor-like. This antibody reacts with human. The C4orf21 / prematurely terminated mRNA decay factor-like Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: C4orf21/prematurely terminated mRNA decay factor-like
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR