Mycoplasma Detection Kit, Southern Biotechnology
Supplier: SOUTHERN BIOTECHNOLOGY MS
13100-01
103259-632EA 283.77 USD
103259-632
Mycoplasma Detection Kit, Southern Biotechnology
Assays Cellular Assays
Mycoplasma Detection Kit for the detection of potential Mycoplasma contamination utilizing PCR.
  • PCR based Mycoplasma detection
  • Detect over 95% of potential cell culture infections
  • Results are typically obtained in less than 3 hours
  • Can detect as little as 2-5 femtograms of Mycoplasma DNA in 100 μL of supernatant
  • Eukaryotic and bacterial DNA are not amplified
SouthernBiotech Mycoplasma Detection Kit was designed to specifically detect potential Mycoplasma contamination in cell cultures. The kit incorporates polymerase chain reaction (PCR) to amplify the conserved 16S ribosomal RNA coding region within the Mycoplasma genome, thereby providing an extensive, highly sensitive and efficient detection method. Primer sequences were carefully determined to cover three genera of Mycoplasmatales (Mycoplasma, Acholeplasma and Ureaplasma) which allows this kit to detect over 95% of potential cell culture infections.
The PCR technology in the Mycoplasma Detection Kit is fast (results are typically obtained in less than 3 hours) and easy to use. The kit is also highly sensitive and can detect as little as 2-5 femtograms of Mycoplasma DNA in 100 µL of test sample supernatant. Eukaryotic and bacterial DNA from cell culture supernatant is not amplified by the kit.
A test cell line infected with Mycoplasma will generate a PCR product(s) between ~448-611-bp on an agarose gel, depending on the type of Mycoplasma present. A positive control (M. orale, 503-bp) has been included to validate that the PCR amplification process has occurred as well as to confirm the size of the PCR product obtained in test samples. An internal control is also provided to eliminate potential false negatives associated with PCR inhibitors.