Alpha1-2,3,6 Mannosidase
Supplier: New England Biolabs (NEB)
P0768S
103258-582EA 159 USD
103258-582
Alpha1-2,3,6 Mannosidase
Enzymes
Recombinant enzyme with no detectable endoglycosidase or other exoglycosidase contaminating activity. 
  • Glycerol-free for optimal performance in HPLC and mass spectrometry analysis
  • ≥95% purity, as determined by SDS-PAGE and intact ESI-MS
  • Optimal activity and stability for up to 12 months
1-2,3,6 Mannosidase, cloned from Jack Bean, and also known as JBM, is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal 1-2, 1-3 and 1-6 linked mannose residues from oligosaccharides. 1-2,3,6 Mannosidase has a slight preference for 1-2 mannose residues over 1-3 and 1-6 mannose residues.