BstZ17I-HF, Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs (NEB)
An E. coli strain that carries the cloned and modified BstZ17I gene from Bacillus stearothermophilus 38M
- Concentration: 20000 units/ml
- 100% activity in CutSmart® buffer
- Time-Saver™ qualified for digestion in 5 to 15 minutes
- Reduced star activity
High-Fidelity (HF) restriction enzymes have the same specificity as native enzymes, but engineered for significantly reduced star activity and performance in a single buffer (CutSmart® Buffer).
Recombinant enzyme with no detectable exoglycosidase or other endoglycosidase contaminating activities.
Delivery information: All HF-restriction enzymes come with gel loading dye, purple (6X). Store at –20 °C.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...