HiFi Taq DNA Ligase
Supplier: New England Biolabs (NEB)
M0647S
103258-566EA 180 USD
103258-566
HiFi Taq DNA Ligase
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
HiFi Taq DNA Ligase is intended for use in molecular diagnostics applications that depend on high fidelity nick ligation
It is not a substitute for T4 DNA ligase and is not suitable for cloning applications or adapter ligation/NGS library preparations.