NEBNext® Multiplex Oligos for Illumina (Index Primers Set 3), New England Biolabs
Supplier: New England Biolabs (NEB)
E7710S E7710L
103258-544EA 116 USD
103258-544 103258-564
NEBNext® Multiplex Oligos for Illumina (Index Primers Set 3), New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.
  • High efficiency adaptor ligation
  • Minimized adaptor-dimer formation
  • Includes index primers for library multiplexing
  • Increased sample identification specificity
  • Includes USER Enzyme