HYPE-5™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
HY01500 HY03000 HY15000 HY30000 HYR10003 HYR10015 HYR20030
103258-460EA 256.83 USD
103258-460 103258-462 103258-464 103258-466 103258-468 103258-470 103258-472
HYPE-5™ Transfection Reagent, OZ Biosciences
Transfection Reagents
HYPE-5 Transfection Kit is dedicated to achieve High Yield Protein Expression in mammalian cells.
  • High yield recombinant protein production
  • Suitable for both HEK293 and CHO cells growing in suspension
  • Compatible with any synthetic or regular media used for protein production
  • Completely animal origin free
This Kit has been designed for maximum recombinant protein expression in HEK293 and CHO cells growing in suspension.
Delivery information: Shipped at room temperature
Packaging: Store Hype-5 Reagent at -20 °C and Hype-5 Blast at +4 °C