ONPG assay kit, OZ Biosciences
Supplier: OZ Biosciences
GO10001
103258-322EA 293.69 USD
103258-322
ONPG assay kit, OZ Biosciences
Assays Enzyme Assays
OZ Biosciences ONPG assay kit is ideal to quantify high expression level of beta-Gal
  • Convenient for all transfection assays
  • Suitable for cultured cells and tissues
  • Simple, rapid and easy-to-use
  • Economical
  • Optimized with stable buffers
All the necessary reagents are provided in this assay kit and offer a rapid, simple and sensitive method to quantify spectrophotometrically the enzyme expression level in LacZ transfected cells.
The levels of active beta-Gal expression can be quickly measured by its catalytic hydrolysis of ONPG (o-nitrophenyl-beta-D-galactopyranoside) substrate to a bright yellow product (Absorbance at 405-420nm).LacZ is one of the most frequently reporter gene used in transfection experiments. The LacZ encoded protein (beta-galactosidase, beta-Gal), is very stable, resistant to proteolytic degradation and easily assayed. 
Delivery information: Shipped with a gel pack
Packaging: Store all reagents at +4°C except Beta Gal Enzyme