In vivo PolyMag™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
In vivo PolyMag Transfection Reagent has been designed for in vivo targeted transfection of various types of nucleic acids such as DNA, RNA, oligonucleotides using magnetic nanoparticles coated with specific polymers.
- Increased transfection efficiency
- Targeted process (magnetically-driven)
- Reduction of the systemic dissemination of vectors during injection
- Reduction of the vector doses
- Work under non permissive conditions (hypothermia, physiological flow conditions)
- Penetration of the vector into tissues
- MInimized toxicity
- Universal - suitable for all nucleic acids
Accessories information: This reagent needs to be used with a magnetic plate.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C