Calcium Phosphate (CaPo) Transfection Kit, OZ Biosciences
Supplier: OZ Biosciences
CP90000
103258-268EA 329.54 USD
103258-268
Calcium Phosphate (CaPo) Transfection Kit, OZ Biosciences
Transfection Reagents
The CaPO transfection kit is simple and easy to use.
  • Ideal for virus production
  • High HEK 293 cells transfection efficiency
  • Suitable for producing recombinant proteins
  • Serum compatible
  • Simple, Ready-to-use and Rapid
It allows reaching between 95 and 100% of HEK 293 transfected cells and a very high titer for virus production.
Delivery information: Shipped at room temperature
Packaging: Store at -20 °C