D-(-)-Luciferin potassium salt ≥99.5% endotoxin-free

Supplier: OZ Biosciences

LK10000
103258-192EA 590.02 USD
103258-192
D-(-)-Luciferin potassium salt ≥99.5% endotoxin-free
D(-)-Luciferin potassium salt

OZ Biosciences D-Luciferin K+ and Na+ salts are routinely used as Firefly’s Luciferase substrate in in vitro and in vivo bioluminescent assays. The quality and purity of the D-Luciferin are essential to obtain good and reproducible results. OZ Biosciences is offering high quality of Endotoxin-Free D-Luciferin K+ and Na+ salts.
Store at -20°C and protect from light


  • High purity > 99.5%
  • Good solubility and great sensitivity
  • Reliable in vivo reporter for bioluminescent assays
  • Endotoxin free (ideal for in vivo application)


Suitable for bioluminescent assays, whole animal imaging, ATP assays, immunoassays, pyrosequencing, luciferase fragment complementation for sequential gene analysis experiments.


Delivery information: Shipped at room temperature

Formula: C₁₁H₇KN₂O₃S₂
MDL Number: MFCD00044928
CAS Number: 115144-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR