pVectOZ™ Transfection Plasmids, OZ Biosciences
Supplier: OZ Biosciences
pVectOZ Transfection Plasmids are engineered in an optimized backbone and are ideal for all transfections.
- Highest levels of transgene expression in mammalian cells and tissues
- Suitable for all transfection applications (in vivo and in vitro)
- High copy number production in Escherichia coli
- LPS-endotoxin free, supercoiled and highly purified: transfection grade approved
OZ Biosciences proposes five expressions vectors encoding for the most popular reporter genes (CAT, GFP, LacZ, Luciferase, SEAP).
Delivery information: Shipped at room temperature
Packaging: Store at -20 °C