Lullaby™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Lullaby is the ideal siRNA transfection reagent for gene silencing
- Effective with very low doses of siRNA
- Minimized off-target effects
- Suitable for siRNA, shRNA, dsRNA, etc.
- Applicable to a broad range of cells
- Serum compatible & non-toxic
- Straightforward protocol
Relying on the TEE-technology, it has been successfully tested on numerous cell lines, reaching up to 90% gene silencing with high reproducibility and a very low toxicity.
Delivery information: Shipped at room temperatue
Packaging: Store at +4 °C