Drum Plug Socket for Round Head, ³/₄", Spark Resistant, Labelmaster
Supplier: Label Master
KSOK75R
103258-076EA 262.02 USD
103258-076
Drum Plug Socket for Round Head, ³/₄", Spark Resistant, Labelmaster
Wrenches
Get the proper fit using ¾" Drum Plug Sockets.
  • Bronze, spark-resistant sockets
  • May be used on round head plugs
  • Fits ⅜" drives on a manual ratchet or manual torque wrench
Be sure to follow the closing instructions for sealing containers in order to be in compliance with UN torque requirements.