Avanti™ J-HC High-Capacity Centrifuge, Beckman Coulter®
Supplier: Beckman Coulter
367501
103255-084EA 62748 USD
103255-084
Avanti™ J-HC High-Capacity Centrifuge, Beckman Coulter®
Centrifuges Floor-Standing Centrifuges
This is a complete solution for batch bioprocessing. Its capacity enables you to process up to nine liters spun at 5000 rpm and 7480×g or 36 liters per hour when harvesting bacteria. And its selection of large-volume swinging bucket and fixed-angle rotors helps accommodate a wide range of processing applications.
Part of a BioSafety system that facilitates rapid harvesting of samples. The samples can be spun directly in this exclusive HarvestLine system liner, which eliminates time-consuming scraping of samples from labware and allow sealing and storing for future use.