ICP-MS Standard, Internal Standard, 6 analytes, Inorganic Ventures
Supplier: INORGANIC VENTURES MS
IV-ICPMS-71D-125
103254-956EA 197.08 USD
103254-956
ICP-MS Standard, Internal Standard, 6 analytes, Inorganic Ventures
Standards Cationic Standards Multi-Element Standards
ICP-MS standard is a certified reference material set in a nitric acid matrix for stability.
6 analytes at 10 µg/ml: Bismuth, Scandium, Yttrium, Indium, Terbium, Lithium-6.
Certifications: Traceable to NIST. Manufactured in accordance with our stingent quality assurance guidelines. ISO 17025 and ISO 17034 certified.