Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: ADVANSTA INC MS
R-05071-500
103254-902EA 143.08 USD
103254-902
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR