Western Blot Strip-it™ Buffer, Advansta
Supplier: ADVANSTA INC MS
R-03722-D50
103254-874EA 169.62 USD
103254-874
Western Blot Strip-it™ Buffer, Advansta
Buffers Electrophoresis Buffers Stripping Buffers
Stripping buffer for stripping antibodies from chemiluminescent Western blots.
  • Conserve valuable samples by probing the same blot multiple times
  • Quickly strip and prepare for reprobing in 25 minutes
  • The gentle protocol removes antibodies without harsh reagents or elevated temperatures
  • Save time by re-probing the same blot
  • Compatible with both nitrocellulose and PVDF membranes and with a variety of chemiluminescent detection reagents
Advansta's Western Blot Strip-It Buffer uniformly removes antibodies from developed Western blots so the blot can be reprobed with another set of antibodies. Stripping and reprobing saves precious samples and the entire stripping protocol takes only 25 minutes. The gentle, odor-free buffer removes bound antibodies without harsh reagents or elevated temperatures and will not damage antigens or membranes.
Conserve your precious sample by reprobing one blot multiple times instead of running duplicate blots. Advansta's Western Blot Strip-It Buffer provides uniform stripping so you can obtain high-quality, qualitative data with multiple probes of the same blot. Strip-It is also compatible with most chemiluminescent reagents.