Anti-IgG Goat Antibody (Dye-490)

Supplier: ADVANSTA INC MS
R-05777-250
103254-852EA 116.54 USD
103254-852
Anti-IgG Goat Antibody (Dye-490)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: 490
Epitope:
Host: Donkey
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR