AdvanBlock™-PF Blocking Solution, Advansta
Supplier: ADVANSTA INC MS
R-03023-D20
103254-748EA 235.38 USD
103254-748
AdvanBlock™-PF Blocking Solution, Advansta
Buffers Electrophoresis Buffers Blocking Buffers
Non-protein blocking solution for fluorescent and chemiluminescent Western blotting
  • Lower background for certain antibodies that require a protein-free solution
  • Optimized to stabilize fluorescence of WesternBright MCF fluorescent secondary antibodies
  • Compatible with fluorescent and chemiluminescent detection
AdvanBlock-PF is a protein-free blocking buffer, optimized for reducing non-specific background in fluorescent and chemiluminescent Western blots. It acts quickly for shorter membrane incubation times, efficiently prevents non-specific binding, and stabilizes the fluorescence of secondary antibody conjugates. AdvanBlock-PF can reduce background when used with primary antibodies that have a high degree of cross-reactivity with protein blockers such as BSA, casein or milk protein. With low-quality primary antibodies that may require protein-based blocking agents, BSA or non-fat dry milk can be dissolved directly in the AdvanBlock-PF solution used to dilute the antibody.
Provided as a 5X concentrate.