Anti-ADAR Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-2168R-HRP
10325-170EA 823.27 USD
10325-170
Anti-ADAR Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
ADAR1 converts adenosine to inosine in dsRNA, which destabilizes the dsRNA helix. This activity is important for various functions like site-specific RNA editing of transcripts of the glutamate receptors and modifying viral RNA genomes (which may be responsible for hypermutation of certain negative-stranded viruses, e.g., measles virus). ADAR1 also binds to short interfering RNAs (siRNA) without editing them and suppresses siRNA-mediated RNA interference. This protein is ubiquitously expressed, with the highest levels being found in brain and lung.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ADAR
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR