Artificial CSF Perfusion Fluid, Harvard Apparatus
Supplier: HARVARD APPARATUS MS
59-7316
103244-114PK 884.62 USD
103244-114
Artificial CSF Perfusion Fluid, Harvard Apparatus
Controls and Calibration Verification Materials
Artificial cerebrospinal fluid (aCSF) is commonly used when sampling from brain interstitial fluid. This solution closely matches the electrolyte concentrations of CSF.
  • Prepared from high purity water and analytical grade reagents
  • Microfiltered and sterile
Final ion concentrations (in mm): Na 150; K 3.0; Ca 1.4; Mg 0.8; P 1.0; Cl 155.
Packaging: Supplied as a package of six 25 ml vials.