CarboFerm™ Neisseria Kit, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z98
10324-438EA
524.61
USD
10324-438
CarboFerm™ Neisseria Kit, Hardy Diagnostics
Microbiology Test Systems
Microorganism Tests
CarboFerm I, rapid carbohydrate test for Neisseria and Moraxella, 24 tests per kit.
For the rapid identification and differentiation of Neisseria spp., including N. gonorrhoeae and N. meningitidis, and Moraxella (Branhamella) catarrhalis. CarboFerm™ uses acid production from carbohydrates to differentiate and identify Neisseria species and M. catarrhalis. Performs a glucose, lactose, maltose, sucrose, and butyrate test. CarboFerm™ Neisseria Kit reduces the time required from the usual 48 hours to as little as 2 - 4 hours!
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)