CatScreen™, Rapid Test for Moraxella (Branhamella) Catarrhalis (M. cat.), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z110
10324-426PK
29.81
USD
10324-426
CatScreen™, Rapid Test for Moraxella (Branhamella) Catarrhalis (M. cat.), Hardy Diagnostics
Microbiology Test Systems
Microorganism Tests
Rapid test for the detection of the enzyme butyrate esterase in bacteria.
- Performs a butyrate test within five minutes
- Presumptive identification of Branhamella (Moraxella) catarrhalis
- Rapid, red color test result
Rapid test for the detection of the enzyme butyrate esterase in bacteria isolated on culture media for the presumptive identification of Branhamella (Moraxella) catarrhalis. Performs a butyrate test within five minutes. Simply moisten disk, inoculate with organism, then wait for the development of a red color.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)