Selective Beta Strep Agar, for Group A Strep only (Streptococcus pyogenes), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
A72
10324-344PK 28.56 USD
10324-344
Selective Beta Strep Agar, for Group A Strep only (Streptococcus pyogenes), Hardy Diagnostics
Cell Culture Supplements and Additives
Preferred for isolating Group A Strep from throat cultures.
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding.
Contains 5% sheep blood, colistin, and trimethoprim sulfamethoxasole (SXT). For the selective isolation of Group A Strep only (S. pyogenes) and exhibits beta hemolysis. Inhibits other Streps, Gram negative bacteria, and normal respiratory flora. Not to be used for group B strep or pneumococcus.
Packaging: Packaged in sleeves of 10 plates in a "breathable" celluose bag that prevents build-up of condensation and excess moisture.