Goat Albumin (from Serum)
Supplier: Equitech-Bio
GSA62-0001 GSA62-0010
103219-920EA 185.77 USD
103219-920 103219-922
Goat Albumin (from Serum)
Proteins and Peptides
Prepared from goat serum by heat-shock fractionation.
  • Purity: ≥ 96%
  • IgG level: not detected
Equitech-Bio offers a wide range of heat-shock and proprietary Cohn method processed animal serum albumins. With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications. Prepared from goat serum by heat-shock fractionation.