Bovine Serum Albumin (BSA)

Supplier: Equitech-Bio

BAH64-0050 BAH64-0100 BAH64-0500 BAH64-1000
103219-852EA 344.08 USD
103219-852 103219-854 103219-856 103219-858
Bovine Serum Albumin (BSA)
Proteins and Peptides

With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications.


  • Reduce shear in bioreactive systems
  • Bind non-specific toxic compounds
  • Serological diluent
  • Molecular weight standard
  • Tissue culture growth nutrient


Equitech-Bio offers a wide range of heat-shock and proprietary Cohn method processed animal serum albumins. With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications. Our albumins are especially useful for creating defined serum-free or serum-reduced mixtures from basal media, reducing shear in bioreactor systems, and binding non-specific toxic compounds. Equitech-Bio offers albumins as a powder as well as a sterile solution. All of Equitech-Bio’s BSA is certified US origin.


Can be used as a serological diluent, as a molecular weight standard, and as a growth nutrient in tissue culture. Tested negative for Blue Tongue and Vesicular Stomatitis.


Ash content: ≤2.0%
Heavy metals: ≤20 ppm
Moisture: ≤5.0%

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR