Bovine Serum Albumin (BSA)

Supplier: Equitech-Bio

BAH62-0100 BAH62-0500 BAH62-1000 BAH63-0100 BAH63-0500 BAH63-1000
103219-840EA 410.71 USD
103219-840 103219-842 103219-844 103219-846 103219-848 103219-850
Bovine Serum Albumin (BSA)
Proteins and Peptides

With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications.


  • Reduce shear in bioreactive systems
  • Bind non-specific toxic compounds
  • Serological diluent
  • Molecular weight standard
  • Tissue culture growth nutrient


Equitech-Bio offers a wide range of heat-shock and proprietary Cohn method processed animal serum albumins. With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications. Our albumins are especially useful for creating defined serum-free or serum-reduced mixtures from basal media, reducing shear in bioreactor systems, and binding non-specific toxic compounds. Equitech-Bio offers albumins as a powder as well as a sterile solution. All of Equitech-Bio’s BSA is certified US origin.


Can be used as a serological diluent, as a molecular weight standard, and as a growth nutrient in tissue culture. Tested negative for Blue Tongue and Vesicular Stomatitis.


Ash content: ≤2.0%
Heavy metals: ≤20 ppm
Moisture: ≤5.0%

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR