Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SGPC35-0100
SGPC35-0500
SGPE35-0100
SGPE35-0500
SGPH35-0100
SGPH35-0500
GPC07-0100
GPC07-0500
GPE07-0100
GPE07-0500
GPH07-0100
103219-068EA
101.27
USD
103219-068
103219-726
103219-728
103219-730
103219-732
103219-734
103219-736
103219-738
103219-740
103219-742
103219-744
103219-746
Goat Plasma, Equitech- Bio, Inc.
Plasma
At Equitech-Bio we maintain our own herd of goats
- Sterility tested
- Endotoxin and mycoplasma testing available
- Lot to lot consistency
Raw plasma is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Sterile-Filtered plasma is sterility tested and sterile-filtered to 0.2 micron absolute. Mycoplasma testing available. Heat-inactivation is available if desired.
Worming histories are controlled and documented.