Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SGPC35-0100 SGPC35-0500 SGPE35-0100 SGPE35-0500 SGPH35-0100 SGPH35-0500 GPC07-0100 GPC07-0500 GPE07-0100 GPE07-0500 GPH07-0100
103219-068EA 101.27 USD
103219-068 103219-726 103219-728 103219-730 103219-732 103219-734 103219-736 103219-738 103219-740 103219-742 103219-744 103219-746
Goat Plasma, Equitech- Bio, Inc.
Plasma
At Equitech-Bio we maintain our own herd of goats
  • Sterility tested
  • Endotoxin and mycoplasma testing available
  • Lot to lot consistency
Raw plasma is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Sterile-Filtered plasma is sterility tested and sterile-filtered to 0.2 micron absolute. Mycoplasma testing available. Heat-inactivation is available if desired.
Worming histories are controlled and documented.