Porcine Serum, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SP30-0100 SP30-0500 PS05-0100 PS05-0500
103219-056EA 96.92 USD
103219-056 103219-058 103219-628 103219-630 103219-632 103219-634
Porcine Serum, Equitech- Bio, Inc.
Sera (Serum)
Collected from animals aged 6 months or older.
  • Sterility tested
  • Endotoxin and mycoplasma testing available
  • Lot to lot consistency
Raw serum is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Serum from specific breeds are available by request. Sterile-Filtered serum is sterility tested and sterile-filtered to 0.2 micron absolute. Endotoxin and mycoplasma testing available. Heat-inactivation is available if desired.
Certifications: Certified U.S. origin.