Feline Serum, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SF30-0100 SF30-0500 FS05--0100 FS05--0500
103219-564EA 670.38 USD
103219-564 103219-566 103219-568 103219-570 103219-572 103219-574
Feline Serum, Equitech- Bio, Inc.
Sera (Serum)
Collected from sexually mature, mixed sex, mixed breed cats.
  • Sterility tested
  • Endotoxin and mycoplasma testing available
  • Lot to lot consistency
Raw serum is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Serum from specific breeds are available by request. Sterile-Filtered serum is sterility tested and sterile-filtered to 0.2 micron absolute. Endotoxin and mycoplasma testing available. Heat-inactivation is available if desired.