Anti-IgG Goat Antibody

Supplier: Equitech-Bio
GAHG80-0100 GAHG80-0500
103219-292EA 369.92 USD
103219-292 103219-294
Anti-IgG Goat Antibody
Antibodies
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope: Heavy and Light Chains
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR