Equine IgG, Equitech- Bio
Supplier: Equitech-Bio
SLE56-00005 SLE56-0001 E60-0001 E60-0010
103219-190EA 155.08 USD
103219-190 103219-192 103219-242 103219-244
Equine IgG, Equitech- Bio
Sera (Serum)
Equitech-Bio's purified immunoglobulin (IgG) preparations are isolated from pooled normal serum by a proprietary fractionation method.
  • Co-precipitant in RIA methods
  • Protein stabilizer
  • Blocker in a number of immunoassays
  • Also useful as IgG control solutions for biochemical analytical tests
Equitech-Bio's purified immunoglobulin (IgG) preparations are isolated from pooled normal serum by a proprietary fractionation method. The immunoglobulin preparations are supplied as a lyophilized powder or liquid solution. Purity levels are >97% IgG as analyzed by SDS-Page. These preparations are especially useful as a co-precipitant in RIA methods employing polyethylene glycol or ammonium sulfate precipitation steps, or as a protein stabilizer or blocker in a number of immunoassays. These preparations are also useful as IgG control solutions for biochemical analytical tests. Immunoglobulin preparations reconstitute to pH 7.2 ± 0.2.