Rat Albumin (from Serum)
Supplier: Equitech-Bio
RTSA62-0050 RTSA62-0100
103219-168EA 575.08 USD
103219-168 103219-170
Rat Albumin (from Serum)
Proteins and Peptides
Prepared from rat serum by heat-shock fractionation.
  • Purity: ≥ 96%
  • IgG level: not detected
Equitech-Bio offers a wide range of heat-shock and proprietary Cohn method processed animal serum albumins. With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications. Prepared from hamster serum by heat-shock fractionation.