Gardner Color Standard #4, Aqua Solutions
Supplier: AQUA SOLUTIONS MS
3914-500ML
103194-014EA
362.97
USD
103194-014
Gardner Color Standard #4, Aqua Solutions
Standards
Color Reference and Spectroscopy Standards
The solution contains potassium platinum chloride, hydrochloric acid 36-38% and water.