Anti-TLR3 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-1444R-CY7
10308-398EA 804.05 USD
10308-398
Anti-TLR3 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Key component of innate and adaptive immunity. TLRs (Toll-like receptors) control host immune response against pathogens through recognition of molecular patterns specific to microorganisms. TLR3 is a nucleotide-sensing TLR which is activated by double-stranded RNA, a sign of viral infection. Acts via the adapter TRIF/TICAM1, leading to NF-kappa-B activation, IRF3 nuclear translocation, cytokine secretion and the inflammatory response (By similarity).

Type: Primary
Antigen: TLR3
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 801-905/905
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR