Wang resin on polystyrene (PS), beads 100-200 mesh Loading: 0.9-1.0 mmol/g

Supplier: Supra Sciences

SPPS36-25 SPPS36-100 SPPS36-1KG
103061-986EA 83.08 USD
103061-986 103061-988 103062-114
Wang resin on polystyrene (PS), beads 100-200 mesh Loading: 0.9-1.0 mmol/g
Wang resin

Common reagent utilized for peptide synthesis on solid support.


Wang Resin is a gel-type polystyrene resin and is a support for solid phase synthesis of peptide amides by coupling amino acids through esterification. This versatile support is capable of forming amines through carbamate formation/cleavage as well as supported small molecule synthesis when carboxylic acids are required. Racemization has been reported utilizing Wang resins, so during activation caution is suggested to minimize the risk. Fmoc protection strategies are commonly employed with Wang and it is customarily cleaved utilizing high percentages of TFA. G

MDL Number: MFCD00131778
CAS Number: 201058-08-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6edc9e14c1d4763ea9a03b28812576bc4694f587&vid=d7da98f0787b59c0b0d7df1c64f7a80c35d1a3fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Colour Off-white
Form Granule
Loading 0.9-1.0 mmol/g
Particle size 100-200 mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR